Kodiak Utility Pressure Gauge, 2 Dia, 1/8" NPT(M) Bottom Connection; 0-600 psi
Representative image only.
Cole-Parmer Mfr # KC101208600 – Item # 6835534
High-quality, low cost gauges designed for industrial processes
  • 1 year warranty
PRODUCT OPTIONS
₹2,800.00INR / Each
₹34,000.00INR / Each
for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy3% from 0 to 200 psi2% from 200 to 400 psi3% from 400 to 600 psi
  • Min Pressure (PSI)0
  • Max Pressure (PSI)600
  • Wetted MaterialsBrass
  • Process Connection1/8" NPT(M)
  • Media CompatibilityGases and liquids compatible with wetted materials
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Utility gauges are designed to replace more expensive digital or commercial gauges