Labconco 7824702 Acid-Resistant 15 x 45 mm Rotor

Representative image only.
Labconco Mfr # 7824702 – Item # 1304097
Enhance the performance of your Labconco CentriVap® Vacuum Concentrator with these accessories
  • Choose replacement parts or optional accessories based on your application needs
  • Accommodates up to 56 auto sample vials 15 x 45 mm
  • Constructed of a thermoplastic for harsh applications

PRODUCT OPTIONS

₹524,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)4
  • Capacity (Gal)56-15 x 45 mm tube
  • MaterialAcid-resistant thermoplastic
  • Compatible WithCentriVap® Centrifugal Vacuum Concentrator systems
  • DescriptionAcid-Resistant 15 x 45 mm Rotor

Key Features

  • Choose replacement parts or optional accessories based on your application needs
  • Accommodates up to 56 auto sample vials 15 x 45 mm
  • Constructed of a thermoplastic for harsh applications

More About this Item

Choose from a wide range of rotors and other accessories suitable for use with your Labconco CentriVap® Complete™ Vacuum Concentrator.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat