Labnet D1125 Block, 6 x 25 mm tubes
Representative image only.
Labnet Mfr # D1125 – Item # 3788512
Uniformly molded high-grade aluminum blocks provide excellent temperature accuracy
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Heaters sold separately
  • PopStopper and AccuRack accessory sold seperately
  • 1 year warranty
PRODUCT OPTIONS
₹28,700.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Length (")3 1/100
  • Width (InchShort)3 35/64
  • Uniformity (° F)0.36
  • Uniformity (° C)0.2
  • Block Number Of Openings6
  • Block - Tube / Vial Size Accepted (mm)25
  • DescriptionLabnet AccuBlock D1125 Block, 6 x 25 mm tubes
  • Warranty1 year
1 Year Warranty
Key Features
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Heaters sold separately
  • PopStopper and AccuRack accessory sold seperately
  • 1 year warranty
More About this Item
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Thermometer wells available in all blocks except those used with plates.