Labnet S2020-P4-220 Microplate Orbital Shaker with Timer, 230 VAC, 50 Hz

Representative image only.
Labnet Mfr # S2020-P4-B – Item # 5140205
Ideal for mixing test solutions in microtitration assays
  • Securely holds four 96-well plates securely with stainless steel spring clamps
  • 1 year warranty

₹292,000.00INR / Each
Consider alternative

SPECS & DESCRIPTION

Specifications & Description

  • Min Speed (rpm)100
  • Max Speed (rpm)1200
  • Platform Length (in)8.5
  • Platform Width (in)7.5
  • Platform Length (cm)21.6
  • Platform Width (cm)19
  • Width (in)6.625
  • Height (in)7.625
  • Depth (in)5.5
  • Width (cm)16.8275
  • Height (cm)19.3675
  • Depth (cm)13.97
  • Max Operating Temperature (° F)149
  • Max Operating Temperature (° C)65
  • TimerYes
  • Timer Range30 seconds to 30 minutes
  • Power (VAC)230
  • Power (Hz)50 Hz
  • Power (amps)0.3
  • DescriptionMicroplate Orbital Shaker with Timer, 230 VAC, 50 Hz
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • Securely holds four 96-well plates securely with stainless steel spring clamps
  • 1 year warranty

More About this Item

Shaker applications include hematology, urology, or EIA/ELISA immunology. Chemical resistanct contoured stainless steel platform holds plates securely.

What''''s included: a platform and a 6-1/4-ft cord with U.S. standard plug

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat