M-System MAKF-120 One-Port Lightning/Surge Protector; Power Supply/120VAC
Representative image only.
M-System Mfr # MAKF-120 – Item # 6846769
Don’t let lightning take systems down
  • Absorb only the lightning surges with no interruption of the instrumentation signal
  • Lightweight, easy-to-handle, plug-in construction
  • Superior protection and selection across a wide range of applications
  • Remove and test without disconnecting wires
  • Base socket connects input-output signals when an element is removed
  • 1 year warranty
PRODUCT OPTIONS
₹31,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 Year
1 Year Warranty
Key Features
  • Absorb only the lightning surges with no interruption of the instrumentation signal
  • Lightweight, easy-to-handle, plug-in construction
  • Superior protection and selection across a wide range of applications
  • Remove and test without disconnecting wires
  • Base socket connects input-output signals when an element is removed