Megger DET2/3 High Performance Ground Tester
Representative image only.
Megger Mfr # 1008-949 – Item # 2000095
Increase safety and performance of electrical systems
  • Obtain measurements in 4P, resistivity, 3P, ART (selective), 2P, and stakeless test modes
  • Operate in the field all day—battery lasts long and recharges quickly
  • Collect high-accuracy earth measurements with 1 mΩ resolution
  • Withstand the elements—IP54 weatherproof with case open, IP65 waterproof when closed
  • Automatic resistance measurements from 0.01 to 200 Ω
  • 1 year warranty
Call For Price
₹34,000.00 - ₹43,100.00INR / Each
for an additional
₹43,100.00INR / Each
for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 Year
1 Year Warranty
Key Features
  • Obtain measurements in 4P, resistivity, 3P, ART (selective), 2P, and stakeless test modes
  • Operate in the field all day—battery lasts long and recharges quickly
  • Collect high-accuracy earth measurements with 1 mΩ resolution
  • Withstand the elements—IP54 weatherproof with case open, IP65 waterproof when closed
  • Automatic resistance measurements from 0.01 to 200 Ω
  • 1 year warranty
More About this Item
This high-performance DET2/3 earth/ground meter provides step-by-step guidance to complete earth tests as required by published standards. Data points are recorded—up to 500 readings—a test report is then built and can be downloaded through USB to a PC. The instrument's battery can last up to 10 hours, and recharges to 100% in three hours—or operate from a standard 12 VDC vehicle power supply.
Order model 20000-96 to obtain wireless remote operation. This tester has additional features to record GPS coordinates, ambient temperature and humidity measurements, and wireless remote control for added safety.
Available for sale in the United States only