Mettler Toledo Expert Go-ISM pH Electrode; BNC with 5.9 ft Cable

Representative image only.
Mettler Toledo Mfr # 51344102 – Item # 3590009
Expertly-engineered probes to cover a wide range of applications
  • ISM® - Intelligent Sensor Management⌡ARGENTHAL™ reference system⌡Open junction works well for dirty and complex samples

PRODUCT OPTIONS

₹194,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • ApplicationsGeneral-use
  • Electrode Body MaterialPEEK
  • Reference JunctionTwo open junctions
  • Reference ElectrolyteNon-refillable Xerolyt® polymer
  • Sensing Glass TypeU - Universal for standard applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • GuardYes
  • ATC ConnectorRCA (cinch)
  • ATC ElementNTC 30 kΩ
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1.8
  • DescriptionpH Electrode; BNC with 5.9 ft Cable

More About this Item

These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.

ISM technology stores calibration data in the sensor and automatically transfer it to the meter. Open junction provides easy cleaning and clog-free measurements. ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat