Mettler Toledo Routine Go-ISM pH Electrode; BNC with 5.9 ft Cable

Representative image only.
Mettler Toledo Mfr # 30248832 – Item # 3590023
Expertly-engineered probes to cover a wide range of applications
  • ISM® - Intelligent Sensor Management⌡ARGENTHAL™ reference system⌡HA glass membrane for high pH and temperature applications

PRODUCT OPTIONS

₹203,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • ApplicationsHigh pH
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference Electrolyte3M KCl
  • Sensing Glass TypeHA - High alkalifor high pH and high temp applications
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature CompensationYes
  • ATC ConnectorRCA (cinch)
  • ATC ElementNTC 30 kΩ
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1.8
  • DescriptionpH Electrode; BNC with 5.9 ft Cable

Key Features

  • ISM® - Intelligent Sensor Management⌡ARGENTHAL™ reference system⌡HA glass membrane for high pH and temperature applications

More About this Item

These ISM® - Intelligent Sensor Management electrodes models store calibration data and automatically transfer the data to your meter. Choose open junction versions for dirty and complex samples. Cables not included with all electrodes. For additional versatility—select the one that connects to your meter.

ISM technology stores calibration data in the sensor and automatically transfer it to the meter. High alkali glass membrane provides low-alkali error—ideal for high pH and high temperature applications. ARGENTHAL™ reference system has a silver ion trap, for silver-ion-free electrolyte. No clogging junction from sulfides or protein binding—perfect for biological samples and TRIS buffers.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat