Oakton 220 pH/ORP/Temperature Controller, 1/8 DIN

Representative image only.
Oakton Item # 5670015
Automatic buffer recognition for quicker setup
  • Designed with both SPDT relay and isolated 4 to 20 mA outputs for controlling and transmitting pH, ORP, and temperature measurements
  • Backlight LCD for clear measurement display
  • Automatic buffer recognition of US and NIST buffers
  • Universal power supply input from 100 to 240VAC
  • Easy-to-install 1/8-DIN compact plastic casing
  • 1 year warranty parts & labor

₹46,000.00INR / Each

₹25,500.00 - ₹34,500.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Control TypeOn/Off
  • Display TypeLCD
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01
  • pH Accuracy±0.1% of reading ±1 digit
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution1
  • mV Accuracy±0.1% of reading ±1 digit
  • Min Temperature (° C)-10
  • Max Temperature (° C)120
  • Min Temperature (° F)14
  • Max Temperature (° F)248
  • Temperature Resolution0.1°C
  • Temperature Accuracy±0.3°C
  • Temperature CompensationAutomatic
  • Output Current4-20mA
  • Output Relay2 SPDT relays
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Width (cm)9.6
  • Height (cm)4.8
  • Depth (cm)11
  • Width (in)3 21/64
  • Height (in)1 3/4
  • Depth (in)4 1/4
  • Panel Mount Size1/8-DIN
  • Warranty1 year parts & labor
1 Year Parts and LaborConformite Europeenne

Key Features

  • Designed with both SPDT relay and isolated 4 to 20 mA outputs for controlling and transmitting pH, ORP, and temperature measurements
  • Backlight LCD for clear measurement display
  • Automatic buffer recognition of US and NIST buffers
  • Universal power supply input from 100 to 240VAC
  • Easy-to-install 1/8-DIN compact plastic casing
  • 1 year warranty parts & labor

More About this Item

Oakton pH/ORP controller features automatic buffer recognition, absolute and relative ORP measurements, and automatic temperature compensation to ensure measurement accuracy. Advanced features include two sets of programmable on/off relays are ideal for high/low control to activate an alarm, a pump, or another device and an isolated 4-20 mA output signal are suitable for opening valves and pacing pumps. ATC element is user selectable, 10K NTC or 1000 Ohm RTD. Order 56105-60 BNC-to-Stripped Wire Adapter for easy connections on terminal strip on back of controller.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat