Oakton ATC probe f/ pH 5, 6, 11, 110, 510, 1100, 2100 and Ion 6 & 510
Representative image only.
Oakton Item # 3561305
Provides ATC for most current Oakton pH meters
  • Ensure optimal pH and ion measurement with ATC probes
  • 6 month warranty
PRODUCT OPTIONS
₹9,500.00INR / Each
pH 5+ Handheld pH Meters
Oakton® pH 5+ Handheld pH Meters
55,000.00 - ₹111,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionATC probe f/ pH 5, 6, 11, 110, 510, 1100, 2100 and Ion 6 & 510
  • Warranty6 month
6 Month Warranty
Key Features
  • Ensure optimal pH and ion measurement with ATC probes
  • 6 month warranty
More About this Item
Corrosion-resistant stainless steel 30 KΩ thermistor features durable polymer handle and 3.3 ft (1-m) cable. Probes measure 88-mm L x 3-mm dia; handle adds another 67 mm to the total length. Temperature range is 0 to 100°C.