Oakton by Cole-Parmer® Submersible 3' L ORP Probe, DJ/ABS/10'/BNC
Representative image only.
Cole-Parmer Item # 2700169
Reach further into solutions
  • Water/wastewater, plating baths
  • Three-foot length for easier measuring in tanks
  • Sturdy design—permanently potted in sturdy ABS plastic housing
  • Submersible up to cable
  • 1 year warranty
PRODUCT OPTIONS
₹73,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialABS plastic
  • Reference JunctionPlatinum
  • Reference ElectrolyteAg/AgCl
  • Max Temperature (° C)80
  • ATC ElementNone
  • ConnectorBNC
  • Diameter (mm)27
  • Length (mm)914.4
  • Length (in)36
  • Cable Length (m)3
  • Warranty1 year
1 Year Warranty
Key Features
  • Water/wastewater, plating baths
  • Three-foot length for easier measuring in tanks
  • Sturdy design—permanently potted in sturdy ABS plastic housing
  • Submersible up to cable
  • 1 year warranty
More About this Item
Submersible electrodes operate from 32 to 176°>F (0 to 80°>C). Submerse up to the cable. Electrodes are potted in a 3 ft L x 1-1/16'' diameter ABS sheath. These electrodes are ideal for measurements in open containers such as tanks or troughs.