Oakton by Cole-Parmer® Submersible 3' L pH Probe, SJ/ABS/100Ohm RTD; Plug/BNC
Representative image only.
Cole-Parmer Item # 0599381
Reach further into solutions
  • Three-foot length for easier measuring in tanks
  • Sturdy design—permanently potted in sturdy ABS plastic housing
  • Submersible up to cable
  • 1 year warranty
PRODUCT OPTIONS
₹67,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsSubmersible
  • Electrode TypeSingle junction
  • Electrode Body MaterialABS plastic
  • Min pH (pH)13
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • ATC ConnectorMini-audio plug
  • ATC Element100 Ω Pt RTD
  • ConnectorBNC
  • Diameter (mm)27
  • Length (mm)914.4
  • Length (in)36
  • Cable Length (m)3
  • Warranty1 year
1 Year Warranty
Key Features
  • Three-foot length for easier measuring in tanks
  • Sturdy design—permanently potted in sturdy ABS plastic housing
  • Submersible up to cable
  • 1 year warranty
More About this Item
Submersible electrodes operate from 32 to 176°>F (0 to 80°>C). Submerse up to the cable. Measure pH from 0 to 13. Electrodes are potted in a 3 ft L x 1-1/16'' diameter ABS sheath. Ideal for measurements in open containers such as tanks or troughs.