Oakton CON 550 Replacement Conductivity Cell K= 1.0
Representative image only.
Oakton Item # 3541932
  • Range of 0 to 200 mS.cm
  • Built-in temperature compensation from 32 to 122°F (0 to 50°C) using a 30KΩ thermistor
  • BNC/RCA connector with 3 ft cable length
  • POM housing material
  • 12 mm diameter and 155 mm length
  • 3 month warranty
PRODUCT OPTIONS
₹21,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionReplacement Conductivity Cell K= 1.0
  • Warranty3 month
3 Month Warranty
Key Features
  • Range of 0 to 200 mS.cm
  • Built-in temperature compensation from 32 to 122°F (0 to 50°C) using a 30KΩ thermistor
  • BNC/RCA connector with 3 ft cable length
  • POM housing material
  • 12 mm diameter and 155 mm length
  • 3 month warranty
More About this Item
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Featured with a proprietary platinum black sensor for accurate readings in a wide conductivity range up to 200 mS/cm, along with a built-in temperature sensor for automatic temperature compensation.