Oakton Electrode Electrolyte Powder
Representative image only.
Oakton Item # 3564070
Optimize water analysis
  • Choose from a variety of accessories
  • Use with Oakton DO probes, 35640-50 & 35640-52
  • 1 year warranty
PRODUCT OPTIONS
₹5,000.00INR / Each
Include Accessories Today!
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithOakton DO probes, 35640-50 & 35640-52
  • DescriptionElectrode Electrolyte Powder
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose from a variety of accessories
  • Use with Oakton DO probes, 35640-50 & 35640-52
  • 1 year warranty
More About this Item
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Add to 500 mL of DI water to make 500 mL of electrolyte