Oakton Flat-Surface Conductivity Probe, K = 1.0, 1 m Cable
Representative image only.
Oakton Item # 3560758
Update, upgrade, or replace to get the best out of your Oakton conductivity, resistivity, or TDS meter
  • Conductivity cells for varying measurement ranges
  • Flat measurement surface allows for readings in sample volumes down to 500 uL
  • Compatible with Oakton CON 11, CON 110, CON 400, and CON 410 meters
  • 1 year warranty
PRODUCT OPTIONS
₹26,200.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithOakton Con 11 & 110 meters
  • Warranty1 year
1 Year Warranty
Key Features
  • Conductivity cells for varying measurement ranges
  • Flat measurement surface allows for readings in sample volumes down to 500 uL
  • Compatible with Oakton CON 11, CON 110, CON 400, and CON 410 meters
  • 1 year warranty
More About this Item
If your conductivity measurements are off, you might need a new cell, sensor, or calibration kit.