Digi-Sense Food Infrared (IR) Thermometer with NIST-Traceable Calibration

Representative image only.
Digi-Sense Item # 3562546
Ideal for food inspections and HACCP programs
  • Non-contact measurement eliminates the risk of cross-contamination
  • Scan numerous surfaces quickly and easily
  • Sealed, hand-washable IP54 housing
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Distance To Target Size Ratio (D:S)2.5:1
  • EmissivityFixed
  • Sighting TypeLED (non-laser)
  • Min Temperature (° F)-20
  • Max Temperature (° F)400
  • Min Temperature (° C)-30
  • Max Temperature (° C)200
  • Accuracy±2.0°F (±1.0°C)
  • Response time<500 milliseconds
  • Battery1 9V (included)
  • Temperature InputYes
  • Warranty1 year
1 Year WarrantyIP54 Rated

Includes

one 9V battery.

Key Features

  • Non-contact measurement eliminates the risk of cross-contamination
  • Scan numerous surfaces quickly and easily
  • Sealed, hand-washable IP54 housing
  • 1 year warranty

More About this Item

Temperature measurement is essential when ensuring proper food storage, cooking and serving environments, and is required to be FDA compliant. Quick checks with a non-contact thermometer can remove the hazard of cross-contamination and reduces cycle time over a traditional penetration probe. This thermometer provides this safe, non-contact reading in under a second.

Use the TempTestr ® to monitor HACCP danger zone temperature standards for food (40 to 140°F or 4 to 60°C), the critical range where harmful bacteria grows most rapidly. LED’s on the meter quickly indicate if the food temperature is within a green or red bacterial growth zone. A green LED light indicates food-safe hot and cold holding temperatures. Red LED light indicates that food is exposed to a potentially dangerous zone.


Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.

For help in selecting an infrared thermometer for industrial, food or laboratory usage please contact the customer service team at Cole-Parmer.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat