Oakton Hygrothermograph Chart Recorder Paper; 14 to 122°F, 7 Day, 2 Hour; 100/Pack
Representative image only.
Oakton Item # 0836821
Replacement paper for Oakton® hygrothermograph chart recorders
  • Varying temperature ranges and speeds
PRODUCT OPTIONS
₹10,100.00INR / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeDrum
  • Chart Speed7 day
  • Chart Width (in)6.625
  • Chart Length (in)11.5
  • Max Temperature (° F)122
  • Min Temperature (° F)14
  • DescriptionHygrothermograph Chart Recorder Paper; 14 to 122°F, 7 Day, 2 Hour; 100/Pack
Key Features
  • Varying temperature ranges and speeds
More About this Item
Choose the paper compatible with your chart recorder and application needs.