Oakton ORP electrode, epoxy body, single junction, 12 mm dia x 110 mm L, 0.9-m cable
Representative image only.
Oakton Item # 3580513
Optimize water analysis
  • Choose from a variety of accessories
  • 1 year warranty
PRODUCT OPTIONS
₹22,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithOakton DO Meter
  • Length (mm)110
  • Diameter (mm)12.5
  • DescriptionORP electrode, epoxy body, single junction, 12 mm dia x 110 mm L, 0.9-m cable
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose from a variety of accessories
  • 1 year warranty
More About this Item
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.