Oakton pH 150 Waterproof Portable Meter Kit with Calibration

Representative image only.
Oakton Item # 3561491
Rugged, accurate, reliable – Best value for an IP67-rated waterproof meter!
  • Intuitive, user-friendly icons—electrode status displays pH slope
  • Grip-Clip™ holder quickly and easily secures the electrode to your beaker or container
  • Expanded data logging for up to 150 data sets
  • mV offset for calibration of ORP standards, zeroing mV value, and pH diagnostics
  • 700 hours battery-life or optional universal power adapter
  • Built-in stand and wall mount option allow for easy benchtop measurement or storage
  • 3 year warranty

PRODUCT OPTIONS

₹152,000.00INR / Each

₹25,500.00 - ₹34,500.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Accessories Today!

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display type3 line custom LCD
  • ConnectionsBNC
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01
  • pH Accuracy±0.01
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 mV for ±999.9 mV; 1 mV beyond ±1000 mV
  • mV Accuracy±0.2
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • WaterproofYes
  • Battery2 AA (included)
  • Battery life (hr)700
  • Width (cm)7.9375
  • Height (cm)5.715
  • Width (in)3 1/8
  • Height (in)2 1/4
  • DescriptionWaterproof Portable Meter Kit with Calibration
  • Warranty3 year
3 Year WarrantyIP 67

Includes

2 AA batteries.

Key Features

  • Intuitive, user-friendly icons—electrode status displays pH slope
  • Grip-Clip™ holder quickly and easily secures the electrode to your beaker or container
  • Expanded data logging for up to 150 data sets
  • mV offset for calibration of ORP standards, zeroing mV value, and pH diagnostics
  • 700 hours battery-life or optional universal power adapter
  • Built-in stand and wall mount option allow for easy benchtop measurement or storage
  • 3 year warranty

More About this Item


What's included: NIST-Traceable calibration certificate provided by InnoCal, All-in-One single-junction pH/ATC electrode 35808-71, Grip-Clip electrode holder, calibration solutions, carrying case, and two AA batteries.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat