Oakton pH Calibration Buffer Pouches, pH 7.00; 20/Pk
Representative image only.
Oakton Item # 3565302
On-the-go convenient pH calibration
  • Single-use pouch—just tear, dip, calibrate
  • Available in common pH values 4, 7, and 10
  • Standardized to NIST-traceable pH references
  • Pack contains 20 pouches
  • NIST/USA buffer value of 7.00 traceable to NIST standard reference materials
  • 1 year warranty
PRODUCT OPTIONS
₹11,100.00INR / Pkg of 20
Need other buffer values?
Oakton® pH Buffer Pouches
11,100.00INR / Pkg of 20
SPECS & DESCRIPTION
Specifications & Description
  • Buffer Kit Values7.00
  • Volume (mL)20
  • DescriptionpH Calibration Buffer Pouches, pH 7.00, 20/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Single-use pouch—just tear, dip, calibrate
  • Available in common pH values 4, 7, and 10
  • Standardized to NIST-traceable pH references
  • Pack contains 20 pouches
  • NIST/USA buffer value of 7.00 traceable to NIST standard reference materials
  • 1 year warranty
More About this Item
Buffers are traceable and offer ±0.01 accuracy at 77 °F (25 °C). Pack of 20 pouches.
pH 7 is a common mid-range pH meter calibration point. All Oakton pH buffer solutions are manufactured under ISO 9000 quality guidelines and traceable to NIST standard (for meters with 0.01 resolution). A pH/temperature table is printed on the label for accurate calibrations.
Certificate of Analysis
Enter lot number
or click to view a Sample C of A