Oakton pH450 Waterproof Portable pH/mV/ISE/Temp Meter with Electrode

Representative image only.
Oakton Item # 3561830
Backlit, rugged waterproof meter is ideal for any environment!
  • Intuitive, user-friendly icons — electrode status displays pH slope
  • Grip-Clip™ electrode holder quickly and easily secures electrode to side of beaker or container
  • USB or RS-232 output for up to 500 data sets
  • Extended battery life or optional universal power adapter
  • Built-in stand with wall mount option allows for easy benchtop measurement or storage
  • 3 year warranty

PRODUCT OPTIONS

₹144,000.00INR / Each

₹25,500.00 - ₹34,500.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Oakton Electrode Storage Solution; 500 mL
₹3,500.00INR / Pint of 1
Include Buffers Today!
Oakton® pH Buffer Solution Calibration Packs
5,100.00 - ₹51,000.00INR / Pkg of 3

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeBacklit LCD
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01
  • pH Accuracy±0.01
  • Buffer RecognitionDIN, NIST, USA, and custom
  • CalibrationUp to 6 points
  • Min mV (mV)-2000
  • Max mV (mV)2000
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±0.2
  • Max Temperature (° C)45
  • InterfaceRS-232 and USB
  • WaterproofYes
  • Battery2 AA (included)
  • Battery life (hr)500
  • Width (cm)7.9375
  • Height (cm)5.715
  • Width (in)3 1/8
  • Length (in)6 1/2
  • Height (in)2 1/4
  • Warranty3 year
3 Year WarrantyConformite EuropeenneIP 67

Includes

2 AA batteries.

Key Features

  • Intuitive, user-friendly icons — electrode status displays pH slope
  • Grip-Clip™ electrode holder quickly and easily secures electrode to side of beaker or container
  • USB or RS-232 output for up to 500 data sets
  • Extended battery life or optional universal power adapter
  • Built-in stand with wall mount option allows for easy benchtop measurement or storage
  • 3 year warranty

More About this Item

Meter includes: "All-in-one" pH/temperature (model 35808-71), Grip-Clip probe holder, and two AA batteries

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat