Oakton pHTestr® 50 Waterproof Pocket pH Tester

Representative image only.
Oakton Item # 3563415
Premium multi parameter pocket testers for field and lab testing
  • Available in pH, Conductivity, TDS, Salinity, and ORP models
  • Replaceable, double-junction sensors for durability and longevity against aggressive samples
  • Four-button keypad for technician level use
  • Leakproof cap functions as a sample cup, hydrating cup, and stabilizing base to stand the meter upright
  • Automatic temperature compensation ensures accurate reading in varying environments
  • 1 year warranty

PRODUCT OPTIONS

₹29,400.00INR / Each

₹17,000.00 - ₹25,900.00INR / Each

for an additional
₹17,000.00INR / Each
for an additional
₹25,900.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Include Buffers Today!
Oakton® pH Buffer Solution Calibration Packs
5,100.00 - ₹51,000.00INR / Pkg of 3

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeTester
  • Display typeLCD
  • Min pH (pH)-1
  • Max pH (pH)15
  • pH Resolution0.01 pH
  • pH Accuracy±0.01 pH
  • Buffer RecognitionUSA 4.01, 7.00, 10.01 and NIST 4.01, 6.86, 9.18
  • Min Temperature (° C)0
  • Max Temperature (° C)60
  • Min Temperature (° F)32
  • Max Temperature (° F)140
  • Temperature Resolution0.1°C / 0.1°F
  • Temperature Accuracy±0.5°C, ±0.9°F
  • Temperature compensationYes, ATC
  • WaterproofYes
  • Battery4 AAA (included)
  • Battery life (hr)250
  • DescriptionWaterproof Pocket pH Tester, Premium 50 Series
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP 67ISO 9001:2008 Certified Supplier

Key Features

  • Available in pH, Conductivity, TDS, Salinity, and ORP models
  • Replaceable, double-junction sensors for durability and longevity against aggressive samples
  • Four-button keypad for technician level use
  • Leakproof cap functions as a sample cup, hydrating cup, and stabilizing base to stand the meter upright
  • Automatic temperature compensation ensures accurate reading in varying environments
  • 1 year warranty

More About this Item

Our most advanced line of pocket testers, the Oakton 50 Series provides single and multimeter options for measuring pH, Conductivity, TDS, Salinity, and ORP. Replaceable double junction sensors provide durability and longevity against highly caustic, volatile or hazardous samples in both lab and field applications. Waterproof and designed to float in water, they are ideal for field-based testing environments – with a convenient loop for security line attachment.

50 series pocket meters include a replaceable double-junction electrode sensor that is designed for more aggressive applications, including wastewater and solutions containing sulfides, heavy metals, and TRIS. The electrode sensor offers a clog-resistant Kynar® junction and a recessed, glass pH bulb to protect against breakage and allow for easy maintenance and replacement. A Valox sensor housing resists most chemicals.

An easy-to-use four-button keypad allows for technician level use and measurement with quick access to a variety of functions. Up to 3 calibration points with auto-buffer recognition at both USA and NIST standard buffers. Additional features include an automatic temperature compensation (ATC), hold function, auto-off function, self-diagnostic error messages, and a three-segment battery-life indicator.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat