Oakton Secondary Standards Kit for Colorimeters
Representative image only.
Oakton Item # 3564570
  • High quality powder reagents for exceptional accuracy, repeatablilty, and stability
  • US-EPA approved DPD method
  • Use secondary standards kit to verify performance of colorimeters for chlorine (free and total), bromine, ozone, and chlorine dioxide.
  • 1 year warranty
PRODUCT OPTIONS
₹36,100.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Test ParametersStandards
  • Warranty1 year
1 Year Warranty
Key Features
  • High quality powder reagents for exceptional accuracy, repeatablilty, and stability
  • US-EPA approved DPD method
  • Use secondary standards kit to verify performance of colorimeters for chlorine (free and total), bromine, ozone, and chlorine dioxide.
  • 1 year warranty
More About this Item
Kit contains one blank and three color reference standards.