Ohaus Dedicated Platform, Stainless Steel, 46 X 46 cm, 500 mL, for use with Reciprocating Shaker

Representative image only.
Ohaus Mfr # 30400081 – Item # 5302746
Expand your reciprocating shaker capabilities
  • 13-500 mL flask clamp capacity
  • 1 year warranty

PRODUCT OPTIONS

₹122,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialStainless Steel
  • Compatible WithOhaus reciprocating shaker
  • Length (InchShort)18
  • Width (in)18
  • Width (cm)46
  • Length (cm)46
  • DescriptionDedicated Platform, Stainless Steel, 46 X 46 cm, 500 mL, for use with Reciprocating Shaker
  • Warranty1 year
1 Year Warranty

Key Features

  • 13-500 mL flask clamp capacity
  • 1 year warranty

More About this Item

Optimize your reciprocating shaker with the following universal, culture, dedicated and funnel platforms. Choose from a variety of platforms sizes to meet your needs. These platforms are available to hold irregular samples or traditional flask clamps, media bottle clamps, and tube racks. Platforms can increase the amount of usable space for your samples, and in some cases can be stacked to increase throughput and double the capacity.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat