Ohaus Dimpled Mat, PVC, 29.8 x 22.2 cm, for use with Waving Shaker SHWV02DG

Representative image only.
Ohaus Mfr # 30400142 – Item # 5302758
Optimize your rocking and waving shakers capabilities
  • 1 year warranty

PRODUCT OPTIONS

₹11,200.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPVC
  • Compatible WithOhaus Waver SHWV02DG
  • Length (InchShort)11.75
  • Width (in)8.75
  • Width (cm)22.2
  • Length (cm)29.8
  • DescriptionDimpled Mat, PVC, 29.8 x 22.2 cm, for use with Waving Shaker SHWV02DG
  • Warranty1 year
1 Year Warranty

Key Features

  • 1 year warranty

More About this Item

Maximize the potential of your rocking and waving shakers with our double-tier platform kits and dimpled mats. The double-tier platforms expand your workspace, allowing you to process more samples simultaneously. Complement these platforms with our dimpled mats, designed to securely hold tubes in place and prevent shifting. This combination not only increases your sample capacity but also enhances efficiency and organization in your laboratory. Upgrade today to optimize your shaker’s performance and streamline your workflow.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat