Ohaus Frontier FC5515R High-Speed Refrigerated Micro Centrifuge with Rotary Knob Control, 15,200 rpm, 120V

Representative image only.
Ohaus Mfr # 30130869 – Item # 8524003
HIgh-speed benchtop micro centrifuges accept a broad range of accessories adding versatility
  • Compact design saves valuable bench space in the lab
  • View actual and set readings on the large backlit LCD
  • Easily maneuver controls even while wearing gloves
  • 10 acceleration/deceleration settings
  • Preset run time from 10 seconds to 99 hours 59 minutes or continuous
  • 99 memory sets to quickly store and recall commonly used parameters
  • Motorized lid-locking system prevents the lid from opening during operation
  • Imbalance sensor stops the rotor when imbalance is detected
  • Refrigerate samples down to -20°C (-4°F)
  • Easy-to-turn rotary knob for quick parameter adjustments
  • High-capacity rotor is ideal for high throughput applications
  • Waterproof keypad provides full protection from spills
  • One-touch toggling between speed, g-force, and temperature controls
  • Powerful induction motor reaches speeds up to 15,200 rpm in less than 16 seconds
  • Maximum RCF of 21,953 x g
  • Biosealed rotor available for safe handling of hazardous samples
  • 2 year warranty

PRODUCT OPTIONS

₹1,497,000.00INR / Each
Often bought with...
Cole-Parmer® ThinTouch Nitrile Gloves, AQL 1.5
530.00 - ₹14,500.00INR / Box of 100

SPECS & DESCRIPTION

Specifications & Description

  • Max Speed (rpm)15200
  • Min Speed (rpm)200
  • Max RCF x g21953
  • Variable SpeedYes
  • Speed ControlDigital, 10 rpm/increment
  • Max Temperature (° C)40
  • Min Temperature (° C)-20
  • Max Temperature (° F)104
  • Min Temperature (° F)-4
  • Temp ControlDigital, 1°C increment
  • RefrigeratedYes
  • DisplayTwo-line, backlit LCD
  • Motor TypeBrushless, induction
  • Noise level (dB(A))≤57
  • Timer Range10 sec to 99 hr 99 min 59 sec or continuous
  • Max Capacity (mL)5
  • Tube Capacity44
  • Length (in)22 1/8
  • Width (in)11 5/16
  • Height (in)11 1/2
  • Length (cm)56.1
  • Width (cm)28.7
  • Height (cm)29.2
  • Power (VAC)120
  • Power (Hz)50/60
  • Power (amps)5.1
  • DescriptionHigh-Speed Refrigerated Micro Centrifuge with Rotary Knob Control, 15,200 rpm, 120V
  • Warranty2 years
Conformite EuropeenneTwo year warranty

Includes

Rotor key and AC power cable.

Key Features

  • Compact design saves valuable bench space in the lab
  • View actual and set readings on the large backlit LCD
  • Easily maneuver controls even while wearing gloves
  • 10 acceleration/deceleration settings
  • Preset run time from 10 seconds to 99 hours 59 minutes or continuous
  • 99 memory sets to quickly store and recall commonly used parameters
  • Motorized lid-locking system prevents the lid from opening during operation
  • Imbalance sensor stops the rotor when imbalance is detected
  • Refrigerate samples down to -20°C (-4°F)
  • Easy-to-turn rotary knob for quick parameter adjustments
  • High-capacity rotor is ideal for high throughput applications
  • Waterproof keypad provides full protection from spills
  • One-touch toggling between speed, g-force, and temperature controls
  • Powerful induction motor reaches speeds up to 15,200 rpm in less than 16 seconds
  • Maximum RCF of 21,953 x g
  • Biosealed rotor available for safe handling of hazardous samples
  • 2 year warranty

More About this Item

The versatile and easy-to-use Frontier benchtop micro centrifuges deliver high speeds, making them ideal for DNA sample preparation, protein separation, and cell pelleting. Quickly make parameter adjustments using a touch wheel or rotary knob. Select from 10 acceleration and 10 deceleration rates to suit your application. Store up to 99 runs with relevant parameters, including the used rotors, to easily repeat the procedures and save time. Safety features include a motorized lid-locking system that prevents the lid from opening during operation and an imbalance sensor that stops the rotor when imbalanced load is detected.

Use the refrigerated model to maintain cool sample temperatures. The centrifuge can maintain temperatures from 40°C down to -20°C (104 to -4 °F) ensuring the integrity of your samples; precool feature quickly lowers the temperature of rotor and chamber before a run. Quickly make parameter adjustments using the easy-to-turn rotary knob; toggle between speed, g-force, and temperature with a single touch. Maintenance-free induction motor produces a maximum RCF of 21,953 x g and reaches speeds up to 15,200 rpm in less than 15 seconds. Use rotor with BIOSEALS when handling hazardous materials. BIOSEALS block the escape of hazardous materials ensuring worker safety.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat