PCRmax PCR Micro Reaction Tube, 0.5 mL
Representative image only.
PCRmax Mfr # 321-05-052 – Item # 6710460
Designed to amplify nucleic acids during the PCR process
  • Manufactured with 99.9% pure virgin polypropylene—withstands temperatures of –20 to 122°>C and are autoclavable
  • Ultra-thin walls accelerate heat transfer providing shorter cycle times
  • Full-length, even insertion cap ensures a tight fit, eliminating sample evaporation
PRODUCT OPTIONS
₹12,400.00INR / Pkg of 1000
SPECS & DESCRIPTION
Specifications & Description
  • Capacity0.5 mL
Key Features
  • Manufactured with 99.9% pure virgin polypropylene—withstands temperatures of –20 to 122°>C and are autoclavable
  • Ultra-thin walls accelerate heat transfer providing shorter cycle times
  • Full-length, even insertion cap ensures a tight fit, eliminating sample evaporation
More About this Item
PCRmax® PCR products are specifically designed for amplifying nucleic acids during the Polymerase Chain Reaction (PCR) process. Certified RNase, DNase, and pyrogen safe to ensure no degradation of the target or amplified product.