PCRmax QPCR KIT QPCR MASTERMIX
Representative image only.
PCRmax Item # 9996666
Quickly and accurately detect the presence of human microbial pathogens
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
PRODUCT OPTIONS
₹43,600.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeDNA
  • Mastermix IncludedYes
  • DescriptionQPCR KIT QPCR MASTERMIX
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.