Plas-Labs Accessory Shelf for Stackable Acrylic Dessicator Cabinets with Gas Ports, Clear, 24" x 18"

Representative image only.
Plas-Labs Mfr # 863-CG.SHF/NS – Item # 0890084
Increase your desiccator's capacity
  • Dimensions: 24" x 18" Color: clear Fits: 08900-62
  • 1 year warranty

PRODUCT OPTIONS

₹21,500.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • DescriptionShelf f/ Stackable Acrylic Dessicator Cabinets w/ Gas Ports, clear, 24" x 18"
  • Warranty1 Year
1 Year Warranty

Key Features

  • Dimensions: 24" x 18" Color: clear Fits: 08900-62
  • 1 year warranty

More About this Item

Add additional shelves to increase your desiccator's capacity.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat