Porta-Lab BS-2001 Microburner Stand with Locking Ring, 4" H
Representative image only.
Porta-Lab Mfr # BS-2001 – Item # 3631050
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)4
  • DescriptionMicroburner Stand with Locking Ring, 4" H
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
These single-ring tripod stands feature a cast-iron top and mild steel legs. The top finished in baked hammertone and the legs are plated and removable.
Tripod ring features three locking grooves cast into the ring for securely holding clay triangles.
Microburner stand for use with microburners. Approximate height is 4'' (10 cm)