Poxygrid F18774-0160 Wire Test Tube Rack, 60, 22-25mm

Representative image only.
Poxygrid Mfr # F18774-0160 – Item # 0673936
Organize and process high volumes of samples
  • 3 month warranty

PRODUCT OPTIONS

₹17,900.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Min Tube Size (mm)22
  • Max Tube Size (mm)25
  • Tubes Held60
  • Length (in)7 3/8
  • Width (in)11 5/8
  • Height (in)3 1/4
  • MaterialSteel
  • DescriptionWire Test Tube Rack, 60, 22-25mm
  • Warranty3 month
3 Month Warranty

Key Features

  • 3 month warranty

More About this Item

Epoxy-coated steel wire racks are chemical and corrosion resistant; unaffected by organic solvents, salts, weak acids, and weak alkalis. Epoxy will withstand a temperature range of -369 to 250°F (-223 to 121°C). Autoclavable

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat