Raven Environmental C-10102 Settlometer Kit, acrylic, 1.4 L, dual scale % solids and SSV
Representative image only.
Raven Environmental Mfr # C-10102 – Item # 0547600
Measure the rate and amount of solid/liquid separation of sludge samples
  • Easy-to-read white calibration markings indicate percent solid and settled sludge volume
  • 3 month warranty
₹25,500.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialAcrylic
  • DescriptionSettlometer Kit, acrylic, 1.4 L, dual scale % solids and SSV
  • Warranty3 month
3 Month Warranty
Key Features
  • Easy-to-read white calibration markings indicate percent solid and settled sludge volume
  • 3 month warranty
More About this Item
This clear plastic settlometer includes mixing paddle, snap-on lid, and instructions. Order the optional interval timer separately below to measure sludge settling rates.