ResinTech UNIX-00 Unix Spring Loaded Single Cartridge Holder, 30 PSI Max
Representative image only.
ResinTech Mfr # UNIX-00 – Item # 9928510
Holds any VP-Series ion exchange water purification cartridge
  • Connect to a pressurized water supply (up to 30 psi)
  • Made in the USA!
  • 1 year warranty
₹47,900.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithResinTech FilterWorks VP-Series Ion Exchange Water Purification Cartridges
  • DescriptionSpring-Loaded Single Ion Exchange Cartridge Holder
  • Warranty1 year
1 Year Warranty
Key Features
  • Connect to a pressurized water supply (up to 30 psi)
  • Made in the USA!
  • 1 year warranty
More About this Item
Holds one cartridge; cartridges may be placed in sequence by mounting multiple holders. Quick, press-to-connect ends for ease of installation.