Saint Gobain Tygopure® Sanitary Coupling, 1/4", Polypropylene
Representative image only.
Saint Gobain Mfr # ATP250PPSB – Item # 5013450
Engineered for a smooth, secure, and contamination-free connection
  • Smooth inner surface minimizes particle entrapment and bacterial growth
  • Leak-proof security ensures reliable tubing connections
  • Tapered design reduces turbulence and stagnation
  • Available in three radiation-sterilizable polymer materials
  • USP Class VI compliant to ensure the highest biopharmaceutical safety standards
PRODUCT OPTIONS
₹10,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolypropylene
  • Sterilization MethodRadiation
  • Connection TypeSanitary coupling to hose barb
  • Barb Size / Tubing Id (Port 1) (in)1/4
  • Sanitary Clamp Size (Port 1) (in)0.985
  • Flow PatternStraight
  • Max Temperature (° F)200
  • Max Temperature (° C)93
  • Fitting Length (in)1 19/32
  • Regulatory ApprovalsUSP
  • WarrantyManufacturer defect
Key Features
  • Smooth inner surface minimizes particle entrapment and bacterial growth
  • Leak-proof security ensures reliable tubing connections
  • Tapered design reduces turbulence and stagnation
  • Available in three radiation-sterilizable polymer materials
  • USP Class VI compliant to ensure the highest biopharmaceutical safety standards
More About this Item
Tygopure® Monobarb Sanitary Fittings are designed for use in bioreactors, filtration systems, media mixing containers, and other critical biopharmaceutical processes. The innovative one-piece monobarb design prevents leaks, reduces contamination risks, and provides a seamless fluid path for gentle and efficient fluid transfer. Available in polypropylene, PVDF, and polysulfone, these fittings are radiation-sterilizable, offering a complete solution for secure and sterile fluid management.