Salisbury by Honeywell 4215 Insulated Universal Switch Stick, 8 foot length, 1.25" diameter
Representative image only.
Salisbury by Honeywell Mfr # 4215 – Item # 8643709
  • 1 year warranty
PRODUCT OPTIONS
₹152,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Length (InchShort)96
  • Diameter (in)1.25
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Also know as hotsticks, these insulated switch sticks are used for opening and closing disconnect switches and cutouts. Also used for installing and removing open-link fuse links. Sticks have a 1.25" (32 mm) diameter and a prong head.