Water Purification MethodAdsorption by spherical activated carbon, catalyst, reverse osmosis, ion exchange, and by point-of-use particle-removing filtration
Max Flow Rate (L/min)1
Inlet Port0.25" OD tubing
With UV LampNo
SourceDirect conection to portable water supply
Type I Water (Resistivity)18.2 MΩ × cm
Feed Water Requirements (Conductivity)< 1500 µS/cm
Feed Water Requirements (TOC)< 2000 ppb
Feed Water Requirements (turbidity)< 1 NTU
Type I Water (Conductivity)0.055 μS/cm
Type I Water (Bacteria)< 0.01 CFU/mL
Feed Water Requirements (pH range)4 to 10
Type I Water (Endotoxins)< 0.001 EU/mL
Feed Water Requirements (pressure)0.5 to 6 bar
Feed Water Requirements (Temperature)2 to 30°C
Type III Water (Bacteria)< 0.01 CFU/mL
Power (VAC)100 to 240
Height (in)20
Depth (in)20.9
Width (in)11
Height (cm)50.9
Depth (cm)53.1
Width (cm)28
DescriptionArium® Mini Plus Starter Set
Warranty2 years
Includes
Arium Mini Plus system, Scientific kit, 5 L bag, and SterilePlus filter.
Key Features
Ideal for small labs requiring <10 L/day of ultrapure water
Large touch display with self-explanatory navigation
Take up a small footprint—only measures 28 cm in width
Volume-controlled dispensing in 50 mL steps
Delivers up to 1 L/minute
Produces ASTM Type I ultrapure water and Type II pure water
Stores water in 5 L Bagtank bag
1 year warranty
More About this Item
Using high-quality water ensures the reproducibility of your results and prevents time-consuming repeat analysis. These compact Arium Mini systems only measure 28 cm (11") wide to easily fit on a benchtop. A high-resolution, touch-activated color display featuring self-explanatory icons is easy to navigate even while wearing gloves. Store favorite functions such repeated dispensing of identical volumes to help increase efficiency while minimizing errors.
Select from Arium Mini, Mini Plus, and Mini Essential depending on your application. All models produce up to 10 L of ASTM Type I ultrapure water per day. Arium Mini and Mini Plus use a closed Bagtank system while Mini Essential has no storage system. For analytical applications, order the Arium Mini with UV lamp to reduce total organic compound (TOC) to = 5 ppb.
Connect Arium Mini Plus water purification system directly to tap water to produce ASTM Type I ultrapure water and Type III pure water. Type I ultrapure water is useful in buffer, media, and reagent preparation and histology, ELISA, AAS, IC and photometry applications. Type III pure water is ideal for general laboratory applications such as rinsing glass and laboratory containers, feed water for laboratory equipment, or feed water for ultrapure water systems.
Arium Mini Plus uses a closed bag tank system which protects pure water from secondary contamination and ensures consistent long-term high water quality. The 5-L bag can be quickly and easily replaced every six months, reducing maintenance time and eliminating the use of hazardous cleaning chemicals.
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.