Sartorius YSTP01 Stat Pen, for Neutralizes Static Charges
Representative image only.
Sartorius Mfr # YSTP01 – Item # 1122967
Helps achieve a more stable and accurate reading by eliminating static
  • 1 year warranty
PRODUCT OPTIONS
₹631,000.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)115
  • DescriptionStat Pen, for Neutralizes Static Charges
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
Eliminate static charge from samples by generating ions with little or no reverse charge—great for highly-electrified samples.