Scienceware H36718-0000 Stainless Steel Vibrating Spatula
Representative image only.
Scienceware Mfr # H36718-0000 – Item # 0626450
Maneuvers easily into small-mouth containers and tight corners
  • Simply spin thumbwheel to control the amount added
  • Ideal for use with analytical balance
PRODUCT OPTIONS
₹23,900.00INR / Pkg of 1
SPECS & DESCRIPTION
Specifications & Description
  • Overall Length (in)8 1/4
  • Overall Length (cm)20.96
  • MaterialStainless Steel
  • SterileNo
  • DescriptionStainless steel vibrating spatula
Key Features
  • Simply spin thumbwheel to control the amount added
  • Ideal for use with analytical balance
More About this Item
Move the toothed thumbwheel to “vibrate” the blade—lets you add as little or as much of your precious material as needed. Narrow V-shaped blade maneuvers easily into small-mouth containers and tight corners. Stainless steel blade is also available with a brown PTFE coating; FDA compliant for direct food contact applications.