Scienceware HDPE Biohazard Bags w/ Sterilization Patch, 12 Gallons, 24 x 30"; 100/CS

Representative image only.
Scienceware Mfr # 13161-0009 – Item # 0649740
Indicator patch darkens when sterile
  • Precautionary procedures printed in four languages
  • Meet/exceed ASTM D 1709-85
  • Autoclavable at 250°F (121°C)

PRODUCT OPTIONS

Loading

₹50,700.00INR / Pkg of 100
Loading

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (Gal)12
  • ColorClear
  • MaterialHDPE
  • Length (in)30
  • Width (in)24
  • Length (cm)76.2
  • Width (cm)60.96
  • Thickness (mil)1.25

More About this Item

These high-molecular weight bags feature an indicator patch that darkens when sterile. Precautionary procedures are printed in English, French, German, and Spanish. Bags are 1.25-mil thick and meet/exceed ASTM D 1709-85.

Optional bag stands are made of aluminum tubing with baked on epoxy coating. The stands fold flat for convenient storage and include a polystyrene safety drip tray that securely supports the bag in the stand. Order stands and plastic ties separately.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat