Scienceware Pipette Support Rack, Holds Up To 50 Pipettes, Polypropylene

Representative image only.
Scienceware Mfr # 189530000 – Item # 0619600
Safe, convenient storage of to 50 pipettes
  • Pipette tips will not contact table surface
  • End plates have handles for easy carrying
  • Steam autoclavable at 121ºC (250ºF)

₹11,400.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Min Tube Size (mm)6
  • Max Tube Size (mm)16
  • Tubes Held50
  • Length (in)8 3/4
  • Width (in)8 3/8
  • Height (in)4 1/2
  • MaterialPolypropylene(PP)
  • TypePipette Support Rack
  • DescriptionPipette Support Rack, Holds Up To 50 Pipettes, Polypropylene

Key Features

  • Pipette tips will not contact table surface
  • End plates have handles for easy carrying
  • Steam autoclavable at 121ºC (250ºF)

More About this Item

Sturdy polypropylene rack is steam autoclavable. Holds up to 50 pipettes in a 5 x 10 arrangement. Top and middle decks have 16 mm diameter holes while bottom deck holes are tapered from 16 mm to 6 mm diameter to prevent pipettes from contacting table, avoiding contamination. Measures 8 3/4" L x 8" W x 4 1/2" H with handles.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat