Spectra Por 131093 Biotech-Grade Dialysis Tubing, 0.5-1.0 kDalton, 24mm, 33ft/roll

Representative image only.
Spectra Por Mfr # 131093 – Item # 0289089
Ideal for concentration, purification, and desalination
  • High-purity membranes with no heavy metal or sulfide contaminants
  • Low protein adsorption
  • 1 year warranty

PRODUCT OPTIONS

₹125,000.00INR / Box of 1

SPECS & DESCRIPTION

Specifications & Description

  • MaterialCellulose Ester, biotech-grade
  • Max Molecular Weight Cut Off (Mwco) (Dalton)1000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)500
  • Flat Width (mm)24
  • Diameter (mm)15
  • Length Per Pack (ft)33
  • Diameter (in)0.591
  • DescriptionBiotech-Grade Dialysis Tubing, 0.5-1.0 kDalton, 24mm, 33ft/roll
  • Warranty1 year
1 Year Warranty

Key Features

  • High-purity membranes with no heavy metal or sulfide contaminants
  • Low protein adsorption
  • 1 year warranty

More About this Item

Cellulose Ester (CE) offers a wider range of more precise MWCO’s for critical separations; packaged as a 10-m roll in 0.05% sodium azide.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat