Spectra Por S/P 7 7 RC Dialysis Tubing, 10000 MWCO; diameter 15 mm; 16 ft (5 m) length.

Representative image only.
Spectra Por Mfr # 132118 – Item # 0289055
Available in a variety of specific MWCO's
  • Ideal for separations sensitive to heavy metal traces
  • 1 year warranty

PRODUCT OPTIONS

₹80,200.00INR / Box of 1

SPECS & DESCRIPTION

Specifications & Description

  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)10000
  • Flat Width (mm)24
  • Diameter (mm)15
  • Length Per Pack (ft)16
  • Diameter (in)0.591
  • Description7 RC Dialysis Tubing, 10000 MWCO; diameter 15 mm; 16 ft (5 m) length.
  • Warranty1 year
1 Year Warranty

Key Features

  • Ideal for separations sensitive to heavy metal traces
  • 1 year warranty

More About this Item

For separations that require specific retentivity and are sensitive to sulfur and metals. EDTA-treated membranes contain no heavy metal traces or sulfur. Supplied wet in 0.1% sodium azide solution. Ready for use after rinsing and conditioning in dialyzing buffer. Each box contains 16-ft of membrane.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat