Supco 6" Circular Chart Recorder Paper, 7 Day, 5 to 45°C; 60/Pack
Representative image only.
Supco Mfr # E545C7 – Item # 8005548
For Supco brand chart recorders
  • Circular and strip charts
  • For CRTH2 and CR4-KTH recorders
  • 1 year warranty
PRODUCT OPTIONS
Loading
₹7,300.00INR / Pkg of 60
Loading
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed7 day
  • Chart Diameter (in)6
  • Max Temperature (° F)113
  • Min Temperature (° F)41
  • Max Temperature (° C)45
  • Min Temperature (° C)5
  • Warranty1 year
1 Year Warranty
More About this Item
Choose the paper compatible with your Supco chart recorder and application needs.