Taylor 5924 Classic Refrigerator / Freezer Analog Extra-Large Dial Thermometer
Representative image only.
Taylor Mfr # 5924 – Item # 9000053
Ensure proper temperature of your food
  • Extra large 3 1/4" dial
  • Temperture range -20 °F to 80 °F and -20 °C to 20 °C
  • Colorful graphics indicate safe temperature zones
  • Red pointer
  • Glass lens
  • Durable stainless steel casing
  • Hangs or stands
  • NSF listed
  • 5 year warranty
PRODUCT OPTIONS
₹1,300.00INR / Each
₹17,000.00 - ₹25,900.00INR / Each
for an additional
₹17,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-20
  • Min Temperature (° C)-28
  • Max Temperature (° F)80
  • Max Temperature (° C)26
  • Dial Size (in)3.25
  • DescriptionRefrigerator / Freezer Analog Extra-Large Dial Thermometer
  • Warranty5 years
5 Year WarrantyNSF international certified
Key Features
  • Extra large 3 1/4" dial
  • Temperture range -20 °F to 80 °F and -20 °C to 20 °C
  • Colorful graphics indicate safe temperature zones
  • Red pointer
  • Glass lens
  • Durable stainless steel casing
  • Hangs or stands
  • NSF listed
  • 5 year warranty
More About this Item
Extra-large Refrigerator/Freezer Dial Thermometer has both a built-in hanger and stand. Dual-scale 3-1/4" (8.3 cm) dia dial shows safe temperature zones. Durable stainless steel construction with a red pointer.