Taylor 5927 TempRite Series Refrigerator / Freezer Glycol-Filled Tube Thermometer
Representative image only.
Taylor Mfr # 5927 – Item # 9000129
Ensure proper temperature of your food
  • Propylene glycol filled tube stimulates food tmperature for more accurate and stable readings.
  • Not affected by changes in ambient air temperature
  • Temperature range -20 to 60°F (-30° to 15°C)
  • NSF Listed
  • Stands, Sits, or Hangs
  • Dimensions: 1 1/4" x 1 1/4" x 5 7/8
  • 1 year warranty
PRODUCT OPTIONS
₹3,100.00INR / Each
₹17,000.00 - ₹25,900.00INR / Each
for an additional
₹17,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-20
  • Min Temperature (° C)-30
  • Max Temperature (° F)60
  • Max Temperature (° C)30
  • Accuracy2%
  • Dial Size (in)3
  • Warranty1 year
1 Year WarrantyNSF international certified
Key Features
  • Propylene glycol filled tube stimulates food tmperature for more accurate and stable readings.
  • Not affected by changes in ambient air temperature
  • Temperature range -20 to 60°F (-30° to 15°C)
  • NSF Listed
  • Stands, Sits, or Hangs
  • Dimensions: 1 1/4" x 1 1/4" x 5 7/8
  • 1 year warranty
More About this Item
TempRite Refrigerator/Freezer Glass Thermometer hangs or stands. Dual scale shows Fahrenheit and Celsius, and includes a safe temperature zone guide. Non-toxic fill solution is safe to clean up in case of breakage. Propylene glycol-filled outer tube simulates food temperature for greater accuracy.