Tektronix Service Solutions DI10102 A2/-4 Intrinsically Safe Ball Bearing Anemometer
Representative image only.
Tektronix Service Solutions Mfr # A2/-4" – Item # 1037459
Accurate measurements in a rugged unit
  • Intrinsically safe 4" mechanical anemometer
  • Accurately measures accumulated air movement and speed
PRODUCT OPTIONS
₹236,000.00INR / Each
₹39,200.00 - ₹49,400.00INR / Each
for an additional
₹39,200.00INR / Each
for an additional
₹49,400.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Air Velocity (ft/min)5000
  • Min Air Velocity (ft/min)30
  • DescriptionA2/-4 Intrinsically Safe Ball Bearing Anemometer
Key Features
  • Intrinsically safe 4" mechanical anemometer
  • Accurately measures accumulated air movement and speed
More About this Item
This first-class hand anemometer has wide appeal, providing the simplest and most reliable measurement of air movement. Rugged and sensitive, yet compact and easy to operate. Used by government departments, mines, industrial plants, laboratories, colleges, contractors, and more. All Davis Instruments anemometers are completely U.S.A. manufactured, assembled and calibrated, and have long life expectancy with lasting accuracy.