Testo 0602 0593 Waterproof Flexible-Tipped Immersion Temperature Probe, Type K TC
Representative image only.
Testo Mfr # 0602 0593 – Item # 3748624
Enhance your temperature monitoring
  • Provide efficient and fast-action measurements
  • 2 year warranty
PRODUCT OPTIONS
₹39,800.00INR / Each
₹34,000.00 - ₹43,100.00INR / Each
for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeType K
  • Min Temperature (° F)-328
  • Max Temperature (° F)1832
  • Min Temperature (° C)-200
  • Max Temperature (° C)1000
  • Response Time (Sec)1
  • Tip Length (in)19.7
  • Tip Diameter (in)0.01
Two year warranty
Includes
Type K mini plug connector and 1.2 m cable.
Key Features
  • Provide efficient and fast-action measurements
  • 2 year warranty
More About this Item
These customized probes are designed to meet your specific needs in monitoring temperatures of food, liquid, and other samples.