Testo 0602 1293 Waterproof Immersion/Penetration Temperature Probe, Type K TC
Representative image only.
Testo Mfr # 0602 1293 – Item # 3748608
Enhance your temperature monitoring
  • Useful for measuring inside liquid wells, pipes, and internal temperature of food
  • 2 year warranty
PRODUCT OPTIONS
₹19,300.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeType K
  • Min Temperature (° F)-76
  • Max Temperature (° F)750
  • Response Time (Sec)7
  • Tip Length (in)4.5
  • Tip Diameter (in)0.02
  • Cable TypeFixed
  • DescriptionWaterproof Immersion/Penetration Temperature Probe, Type K TC
Two year warranty
Key Features
  • Useful for measuring inside liquid wells, pipes, and internal temperature of food
  • 2 year warranty
More About this Item
These customized probes are designed to meet your specific needs in monitoring temperatures of food, liquid, and other samples.