Testo 0602 4592 Pipe Clamp Temperature Probe, Type K, 1/5" to 2-1/2"
Representative image only.
Testo Mfr # 0602 4592 – Item # 3748619
Enhance your temperature monitoring
  • Pipe wrap probe for pipe diameter 1/5" to 2-1/2"
  • Measurement range short-term to 536°F
  • 2 year warranty
PRODUCT OPTIONS
₹55,900.00INR / Each
₹34,000.00 - ₹43,100.00INR / Each
for an additional
₹34,000.00INR / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeType K
  • Min Temperature (° F)-76
  • Max Temperature (° F)266
  • Response Time (Sec)5
  • Cable TypeFixed
  • Cable Length (in)47 1/4
  • DescriptionPipe Clamp Temperature Probe, Type K; 1/5" to 2 1/2"
Two year warranty
Key Features
  • Pipe wrap probe for pipe diameter 1/5" to 2-1/2"
  • Measurement range short-term to 536°F
  • 2 year warranty
More About this Item
These customized probes are designed to meet your specific needs in monitoring temperatures of food, liquid, and other samples.