Texwipe TX752E ESD-Safe CleanFoam® Medium Compressed Cleanroom Swabs, Non-Sterile; 500/bag.

Representative image only.
Texwipe Mfr # TX752E – Item # 3367621
Eliminates risk of adhesive contamination
  • Thermal bond construction
  • No blooming carbon particles, metals, or ionic antistatic contaminants
  • Lot coded for traceability and quality control
  • Certified to ISO 9001:2015

PRODUCT OPTIONS

₹30,800.00INR / Bag of 500

SPECS & DESCRIPTION

Specifications & Description

  • Handle Length (in)3.169
  • ComplianceISO 9001:2015
  • Tip Length (InchShort)25/64
  • Tip Length (cm)1
  • Swab Total Length (InchShort)3 9/16
  • Swab Total Length (cm)9.05

Key Features

  • Thermal bond construction
  • No blooming carbon particles, metals, or ionic antistatic contaminants
  • Lot coded for traceability and quality control
  • Certified to ISO 9001:2015

More About this Item

The ESD-Safe cleanroom swabs feature Stat-Rite® handle, a patented dissipative polymer for clean ESD protection without contributing particulate contaminants. The head material is thermally bonded to the handle to eliminate the risk of adhesive contamination.

Easily apply and remove lubricants, adhesives, and other solutions in a critical clean environment with EDS concerns. The swabs are also chemical compatibility with a variety of solutions and solvents.

Cleanroom processed, the swabs are packaged in an anti-static bag. Packaging includes 500 swabs/bag, 5 inner bags of 100 swabs, 5 bags/case for a total 2,500 swabs/case. Choose from CleanFoam®, Alpha® polyester kit, or dual micro-tip pick.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat